Learn More
Androgen Receptor Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and gel supershift experiments.
Brand: Epredia MS-443-P
Code : LD
Additional Details : Weight : 4.98950kg
Description
The expression of AR is reportedly inversely correlated with histologic grade, i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed as a marker of hormone-responsiveness.Host Species: Mouse
Clone: AR 441
Isotype: IgG1
Species Reactivity: Human and Dog. Does not react with mouse. Others not tested.
Epitope: aa 299-315
Immunogen: A synthetic peptide from human AR
Molecular Weight: 110kDa
Positive Control: LnCAp cells or prostate CA
Cellular Localization: Nuclear
Ab-1 is excellent for staining of formalin/paraffin tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifications
Androgen Receptor Ab-1 | |
Monoclonal | |
200μg/mL | |
Mouse | |
1 mL | |
Steroids Hormones and Growth Factors | |
Canine, Human | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
AR 441 | |
Unconjugated | |
A synthetic peptide from human AR | |
RUO | |
Primary | |
Concentrated |