missing translation for 'onlineSavingsMsg'
Learn More

JMJD2B Antibody (3A6H6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£161.00 - £435.00

Specifications

Antigen JMJD2B
Clone 3A6H6
Concentration 1 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Applications Western Blot, Flow Cytometry, ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18238172
View Documents
Novus Biologicals
NBP2-61800
0.1 mL
£435.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18613048
View Documents
Novus Biologicals
NBP2-61800-0.025ml
0.025 mL
£161.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

JMJD2B Monoclonal specifically detects JMJD2B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

JMJD2B
1 mg/mL
Western Blot, Flow Cytometry, ELISA
Unconjugated
Mouse
Human, Mouse
23030
Purified recombinant fragment of human KDM4B (AA: 960-1096) expressed in E. Coli.
Primary
122 kDa
3A6H6
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Monoclonal
Purified
Chromatin Modifiers, Chromatin Research, Epigenetics
EC 1.14.11, FLJ44906, JHDM3B, JmjC domain-containing histone demethylation protein 3B, JMJD2BEC 1.14.11.-, jumonji domain containing 2B, Jumonji domain-containing protein 2B, KIAA0876lysine-specific demethylase 4B, lysine (K)-specific demethylase 4B
KDM4B
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.