missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD2B Antibody (3A6H6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61800
This item is not returnable.
View return policy
Description
JMJD2B Monoclonal specifically detects JMJD2B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| JMJD2B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| EC 1.14.11, FLJ44906, JHDM3B, JmjC domain-containing histone demethylation protein 3B, JMJD2BEC 1.14.11.-, jumonji domain containing 2B, Jumonji domain-containing protein 2B, KIAA0876lysine-specific demethylase 4B, lysine (K)-specific demethylase 4B | |
| Mouse | |
| 122 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 3A6H6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| KDM4B | |
| Purified recombinant fragment of human KDM4B (AA: 960-1096) expressed in E. Coli. | |
| Protein G purified | |
| Chromatin Modifiers, Chromatin Research, Epigenetics | |
| 23030 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction