missing translation for 'onlineSavingsMsg'
Learn More

JMJD2B Antibody (3A6H6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-61800-0.025ml

 View more versions of this product

Product Code. 18613048

  • £161.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

JMJD2B Monoclonal specifically detects JMJD2B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

JMJD2B
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
EC 1.14.11, FLJ44906, JHDM3B, JmjC domain-containing histone demethylation protein 3B, JMJD2BEC 1.14.11.-, jumonji domain containing 2B, Jumonji domain-containing protein 2B, KIAA0876lysine-specific demethylase 4B, lysine (K)-specific demethylase 4B
Mouse
122 kDa
0.025 mL
Primary
Human, Mouse
Purified
Western Blot, Flow Cytometry, ELISA
3A6H6
Unconjugated
PBS with 0.05% Sodium Azide
KDM4B
Purified recombinant fragment of human KDM4B (AA: 960-1096) expressed in E. Coli.
Protein G purified
Chromatin Modifiers, Chromatin Research, Epigenetics
23030
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.