missing translation for 'onlineSavingsMsg'
Learn More

Jarid1A Rabbit anti-Human, Mouse, Rat, Clone: 5B10P1, Novus Biologicals™

Rabbit Monoclonal Antibody

£158.00 - £387.00

Specifications

Antigen Jarid1A
Clone 5B10P1
Dilution Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200
Applications ChIP Assay, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18312114
View Documents
Novus Biologicals
NBP3-16478-20UL
20 μg
£158.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18013404
View Documents
Novus Biologicals
NBP3-16478-100UL
100 μg
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Jarid1A Monoclonal antibody specifically detects Jarid1A in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunoprecipitation
Specifications

Specifications

Jarid1A
Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1A, JARID1AJumonji, AT rich interactive domain 1A (RBP2-like), Jumonji, AT rich interactive domain 1A (RBBP2-like), Jumonji/ARID domain-containing protein 1A, KDM5A, lysine (K)-specific demethylase 5A, RBBP-2, RBBP2lysine-specific demethylase 5A, RBP2retinoblastoma binding protein 2, Retinoblastoma-binding protein 2jumonji, AT rich interactive domain 1A
Recombinant fusion protein containing a sequence corresponding to amino acids 1400-1690 of human Jarid1A (P29375). HAYSSASKSCSQGSSTPRKQPRKSPLVPRSLEPPVLELSPGAKAQLEELMMVGDLLEVSLDETQHIWRILQATHPPSEDRFLHIMEDDSMEEKPLKVKGKDSSEKKRKRKLEKVEQLFGEGKQKSKELKKMDKPRKKKLKLGADKSKELNKLAKKLAKEEERKKKKEKAAAAKVELVKESTEKKREKKVLDIPSKYDWSGAEESDDENAVCAAQNCQRPCKDKVDWVQCDGGCDEWFHQVCVGVSPEMAENEDYICINCAKKQGPVSPGPAPPPSFIMSYKLPMEDLKETS
Primary
Store at -20°C. Avoid freeze-thaw cycles.
5B10P1
ChIP Assay, Immunoprecipitation
Unconjugated
Rabbit
Cell Biology
PBS, 0.05% BSA, 50% glycerol, pH7.3
5927
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.