missing translation for 'onlineSavingsMsg'
Learn More

Jarid1A Rabbit anti-Human, Mouse, Rat, Clone: 5B10P1, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-16478-20UL

 View more versions of this product

Product Code. 18312114

  • £158.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Jarid1A Monoclonal antibody specifically detects Jarid1A in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Jarid1A
Monoclonal
Unconjugated
PBS, 0.05% BSA, 50% glycerol, pH7.3
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
ChIP Assay, Immunoprecipitation
5B10P1
Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200
EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1A, JARID1AJumonji, AT rich interactive domain 1A (RBP2-like), Jumonji, AT rich interactive domain 1A (RBBP2-like), Jumonji/ARID domain-containing protein 1A, KDM5A, lysine (K)-specific demethylase 5A, RBBP-2, RBBP2lysine-specific demethylase 5A, RBP2retinoblastoma binding protein 2, Retinoblastoma-binding protein 2jumonji, AT rich interactive domain 1A
Recombinant fusion protein containing a sequence corresponding to amino acids 1400-1690 of human Jarid1A (P29375). HAYSSASKSCSQGSSTPRKQPRKSPLVPRSLEPPVLELSPGAKAQLEELMMVGDLLEVSLDETQHIWRILQATHPPSEDRFLHIMEDDSMEEKPLKVKGKDSSEKKRKRKLEKVEQLFGEGKQKSKELKKMDKPRKKKLKLGADKSKELNKLAKKLAKEEERKKKKEKAAAAKVELVKESTEKKREKKVLDIPSKYDWSGAEESDDENAVCAAQNCQRPCKDKVDWVQCDGGCDEWFHQVCVGVSPEMAENEDYICINCAKKQGPVSPGPAPPPSFIMSYKLPMEDLKETS
20 μg
Cell Biology
5927
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.