missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jarid1A Rabbit anti-Human, Mouse, Rat, Clone: 5B10P1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-16478-100UL
This item is not returnable.
View return policy
Description
Jarid1A Monoclonal antibody specifically detects Jarid1A in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunoprecipitation
Specifications
| Jarid1A | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ChIP Assay, Immunoprecipitation | |
| 5B10P1 | |
| Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200 | |
| EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1A, JARID1AJumonji, AT rich interactive domain 1A (RBP2-like), Jumonji, AT rich interactive domain 1A (RBBP2-like), Jumonji/ARID domain-containing protein 1A, KDM5A, lysine (K)-specific demethylase 5A, RBBP-2, RBBP2lysine-specific demethylase 5A, RBP2retinoblastoma binding protein 2, Retinoblastoma-binding protein 2jumonji, AT rich interactive domain 1A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1400-1690 of human Jarid1A (P29375). HAYSSASKSCSQGSSTPRKQPRKSPLVPRSLEPPVLELSPGAKAQLEELMMVGDLLEVSLDETQHIWRILQATHPPSEDRFLHIMEDDSMEEKPLKVKGKDSSEKKRKRKLEKVEQLFGEGKQKSKELKKMDKPRKKKLKLGADKSKELNKLAKKLAKEEERKKKKEKAAAAKVELVKESTEKKREKKVLDIPSKYDWSGAEESDDENAVCAAQNCQRPCKDKVDWVQCDGGCDEWFHQVCVGVSPEMAENEDYICINCAKKQGPVSPGPAPPPSFIMSYKLPMEDLKETS | |
| 100 μg | |
| Cell Biology | |
| 5927 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction