Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps is a gas collecting tube with O-ring valves on each end.
£114.00 - £148.00
Product Code | Brand | Measurement Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Measurement Range | Price | Quantity & Availability | |||||
15834774
|
GPE Scientific™
GST/100 |
100 mL |
£114.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
15854774
|
GPE Scientific™
GST/125 |
125 mL |
£115.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
15874774
|
GPE Scientific™
GST/250 |
250 mL |
£136.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
15884774
|
GPE Scientific™
GST/500 |
500 mL |
£123.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
15844774
|
GPE Scientific™
GST/1000 |
1000 mL |
£143.00
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
15864774
|
GPE Scientific™
GST/2000 |
2000 mL |
£148.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
Description
- Cylindrical, oblong shape
- Gas tight container
- Gauged volume
- Standard collecting tube with valved ends
Specifications
Gas Sample Tube |