Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps
Product Code. 15884774
Click to view available options
Measurement Range:
100 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Product Code. 15884774
Brand: GPE Scientific™ GST/500
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps is a gas collecting tube with O-ring valves on each end.
  • Cylindrical, oblong shape
  • Gas tight container
  • Gauged volume
  • Standard collecting tube with valved ends
Specifications
Type Gas Sample Tube
Measurement Range 500 mL