Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps is a gas collecting tube with O-ring valves on each end.
Brand:  GPE Scientific™ GST/100
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15834774
  • £114.00 / Each
Estimated Shipment: 19-07-2024
to see stock.
Add to basket
Description
Description
  • Cylindrical, oblong shape
  • Gas tight container
  • Gauged volume
  • Standard collecting tube with valved ends
Specifications
Specifications
Gas Sample Tube
100 mL