missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRS1, p Ser639 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£193.00 - £320.00
Specifications
| Antigen | IRS1 (p Ser639) |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18235564
|
Novus Biologicals
NB100-82000-0.025ML |
0.025 mL |
£193.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109369
|
Novus Biologicals
NB100-82000 |
0.05 mL |
£320.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRS1 Polyclonal specifically detects IRS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IRS1 (p Ser639) | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| HIRS-1, insulin receptor substrate 1, IRS-1 | |
| IRS1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Autophagy, Cancer, Diabetes Research, mTOR Pathway, Nutrient Sensing in the Brain, Phospho Specific, Signal Transduction | |
| P35568 | |
| 3667 | |
| IgG | |
| Affinity Purified | |
| 180 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title