missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRS1, p Ser639 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-82000
This item is not returnable.
View return policy
Description
IRS1 Polyclonal specifically detects IRS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IRS1 (p Ser639) | |
| Polyclonal | |
| Unconjugated | |
| PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide | |
| HIRS-1, insulin receptor substrate 1, IRS-1 | |
| Rabbit | |
| 180 kDa | |
| 0.05 mL | |
| Autophagy, Cancer, Diabetes Research, mTOR Pathway, Nutrient Sensing in the Brain, Phospho Specific, Signal Transduction | |
| 3667 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| P35568 | |
| IRS1 | |
| The antiserum was produced against synthesized phosphopeptide derived from human IRS1 around the phosphorylation site of serine 639 (P-K-SP-V-S). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects endogenous levels of IRS1 only when phosphorylated at serine 639. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction