missing translation for 'onlineSavingsMsg'
Learn More

IRS1, p Ser639 Antibody, Novus Biologicals™

Product Code. 18235564 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
Unit Size:
0.05mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18235564 0.025 mL 25µL
18109369 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18235564 Supplier Novus Biologicals Supplier No. NB100820000.025ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IRS1 Polyclonal specifically detects IRS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRS1 (p Ser639)
Applications Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS [pH 7.4] 150mM NaCl, and 50% glycerol (without Mg2+ and Ca2+) with 0.02% Sodium Azide
Gene Accession No. P35568
Gene Alias HIRS-1, insulin receptor substrate 1, IRS-1
Gene Symbols IRS1
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human IRS-1 around the phosphorylation site of serine 639 (P-K-SP-V-S).
Molecular Weight of Antigen 180 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Diabetes Research, mTOR Pathway, Nutrient Sensing in the Brain, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3667
Test Specificity IRS-1 (specific to Phospho-Ser639) detects endogenous levels of IRS-1 only when phosphorylated at serine 639.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Vis mindre

For Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.