missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Wash Buffer (25X)
Suitable for use with all ELISA assays
£33.58 - £109.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10334443
|
Invitrogen™
WB01 |
100 mL |
£33.58
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
10140523
|
Invitrogen™
WB02 |
1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Clear, colorless concentratred liquid buffer
- Available in two sizes
- Detection method: Colormetric
Build Your Own Immunoassay, Cell Analysis, Immunoassays
Order Info
Shipping Condition: Wet Ice
Specifications
Wet Ice |