missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Wash Buffer (25X)

Product Code. 10334443
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10334443 100 mL Each
10140523 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10334443 Supplier Invitrogen™ Supplier No. WB01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Suitable for use with all ELISA assays

  • Clear, colorless concentratred liquid buffer
  • Available in two sizes
  • Detection method: Colormetric

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Content And Storage Buffer. Store at 2°C to 8°C.
Shipping Condition Wet Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.