Learn More
Agilent BioTek Imaging Objectives for Lionheart FX Automated Live Cell Imager
For use with Lionheart FX Automated Live Cell Imager.
Brand: Agilent BioTek 1450539
Description
- Includes oil and absorbing ring
Specifications
100X Oil Objective Olympus, 1.40 NA | |
Lionheart FX Automated Live Cell Imager |