missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-26/AK155 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
POA
Specifications
Antigen | IL-26/AK155 |
---|---|
Applications | Western Blot |
Conjugate | Unconjugated |
Host Species | Goat |
Regulatory Status | RUO |
Description
IL-26/AK155 Polyclonal specifically detects IL-26/AK155 in Human samples. It is validated for Western Blot.Specifications
IL-26/AK155 | |
Unconjugated | |
RUO | |
55801 | |
Polyclonal | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
Goat | |
Human | |
E. coli-derived recombinant human IL-26 (R&D Systems, Catalog # 1375-IL) Lys22-Gln171 Accession # Q9NPH9 | |
Primary |
For Research Use Only