missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-26/AK155 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1375
This item is not returnable.
View return policy
Description
IL-26/AK155 Polyclonal specifically detects IL-26/AK155 in Human samples. It is validated for Western Blot.
Specifications
| IL-26/AK155 | |
| Polyclonal | |
| Unconjugated | |
| Q9NPH9 | |
| IL26 | |
| E. coli-derived recombinant human IL-26 (R&D Systems, Catalog # 1375-IL) Lys22-Gln171 Accession # Q9NPH9 | |
| 100 μg | |
| Primary | |
| Detects human IL-26 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human IL-10 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AK155, AK155IL-26interleukin-26, IL26, interleukin 26, Protein AK155 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55801 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction