missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-26/AK155 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF1375-SP
This item is not returnable.
View return policy
Description
IL-26/AK155 Polyclonal specifically detects IL-26/AK155 in Human samples. It is validated for Western Blot.
Specifications
| IL-26/AK155 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AK155, AK155IL-26interleukin-26, IL26, interleukin 26, Protein AK155 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55801 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9NPH9 | |
| IL26 | |
| E. coli-derived recombinant human IL-26 (R&D Systems, Catalog # 1375-IL) Lys22-Gln171 Accession # Q9NPH9 | |
| 25 μg | |
| Primary | |
| Detects human IL-26 in direct ELISAs and Western blots. In these formats, less than 5% cross-reactivity with recombinant human IL-10 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction