missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ikaros/IKZF1 Antibody (21E9), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Ikaros/IKZF1 |
|---|---|
| Clone | 21E9 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30036376
|
Novus Biologicals
NBP3-26204-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30033845
|
Novus Biologicals
NBP3-26204-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ikaros/IKZF1 Monoclonal antibody specifically detects Ikaros/IKZF1 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Ikaros/IKZF1 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DNA-binding protein Ikaros, hIk-1, IK1LyF-1, Ikaros (zinc finger protein), IKAROS family zinc finger 1 (Ikaros), Ikaros family zinc finger protein 1, IKAROSLymphoid transcription factor LyF-1, LYF1PRO0758, zinc finger protein, subfamily 1A, 1 (Ikaros), ZNFN1A1CLL-associated antigen KW-6 | |
| A synthesized peptide derived from Human Ikaros/IKZF1 [UniProt Q13422] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 21E9 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Innate Immunity | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 10320 | |
| IgG | |
| Affinity purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts