missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ikaros/IKZF1 Antibody (21E9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26204-50ul
This item is not returnable.
View return policy
Description
Ikaros/IKZF1 Monoclonal antibody specifically detects Ikaros/IKZF1 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| Ikaros/IKZF1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 21E9 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| DNA-binding protein Ikaros, hIk-1, IK1LyF-1, Ikaros (zinc finger protein), IKAROS family zinc finger 1 (Ikaros), Ikaros family zinc finger protein 1, IKAROSLymphoid transcription factor LyF-1, LYF1PRO0758, zinc finger protein, subfamily 1A, 1 (Ikaros), ZNFN1A1CLL-associated antigen KW-6 | |
| A synthesized peptide derived from Human Ikaros/IKZF1 [UniProt Q13422] | |
| 50 μL | |
| Innate Immunity | |
| 10320 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction