missing translation for 'onlineSavingsMsg'
Learn More

Ikaros/IKZF1 Antibody (21E9), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-26204-100ul

 View more versions of this product

Product Code. 30033845

  • £406.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ikaros/IKZF1 Monoclonal antibody specifically detects Ikaros/IKZF1 in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ikaros/IKZF1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA
21E9
Flow Cytometry 1:50-1:200, ELISA
DNA-binding protein Ikaros, hIk-1, IK1LyF-1, Ikaros (zinc finger protein), IKAROS family zinc finger 1 (Ikaros), Ikaros family zinc finger protein 1, IKAROSLymphoid transcription factor LyF-1, LYF1PRO0758, zinc finger protein, subfamily 1A, 1 (Ikaros), ZNFN1A1CLL-associated antigen KW-6
A synthesized peptide derived from Human Ikaros/IKZF1 [UniProt Q13422]
100 μL
Innate Immunity
10320
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.