missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT88 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | IFT88 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16872414
|
Proteintech
13967-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16862414
|
Proteintech
13967-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody was raised against the C-terminal region of human IFT88 and can detect the endogenous level of IFT88.
This gene encodes a member of the tetratrico peptide repeat family. Mutations of a similar gene in mouse can cause polycystic kidney disease. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
| IFT88 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Canine, Human | |
| Q13099, Q61371 | |
| 21821, 305918, 477345, 8100 | |
| IFT88 Fusion Protein Ag4980 | |
| Primary | |
| -20°C | |
| IFT88 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| IFT88, TPR repeat protein 10, TTC10 | |
| IFT88 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title