missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT88 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13967-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody was raised against the C-terminal region of human IFT88 and can detect the endogenous level of IFT88.
This gene encodes a member of the tetratrico peptide repeat family. Mutations of a similar gene in mouse can cause polycystic kidney disease. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
| IFT88 | |
| Polyclonal | |
| Unconjugated | |
| IFT88 | |
| IFT88, TPR repeat protein 10, TTC10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21821, 305918, 477345, 8100 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13099, Q61371 | |
| IFT88 | |
| IFT88 Fusion Protein Ag4980 | |
| 150 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction