missing translation for 'onlineSavingsMsg'
Learn More

IFT88 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16872414 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16872414 20 μL 20µL
16862414 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16872414

Brand: Proteintech 139671AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody was raised against the C-terminal region of human IFT88 and can detect the endogenous level of IFT88.

This gene encodes a member of the tetratrico peptide repeat family. Mutations of a similar gene in mouse can cause polycystic kidney disease. Two transcript variants encoding distinct isoforms have been identified for this gene.

Specifications

Antigen IFT88
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IFT88
Gene Accession No. Q13099, Q61371
Gene Alias IFT88, TPR repeat protein 10, TTC10
Gene Symbols IFT88
Host Species Rabbit
Immunogen IFT88 Fusion Protein Ag4980
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21821, 305918, 477345, 8100
Target Species Rat, Mouse, Canine, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.