missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human XCR1 PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 1 publication
£196.00 - £442.00
Specifications
| Antigen | XCR1/CCXCR1 |
|---|---|
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | R-PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18752229
![]() |
R&D Systems
FAB857P-025 |
25 Tests |
£196.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18787786
![]() |
R&D Systems
FAB857P-100 |
100 Tests |
£442.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
XCR1/CCXCR1 Polyclonal specifically detects XCR1/CCXCR1 in Human samples. It is validated for Flow Cytometry.Specifications
| XCR1/CCXCR1 | |
| Flow Cytometry | |
| R-PE | |
| Human | |
| CCXCR1, CCXCR1XC chemokine receptor 1, chemokine (C motif) receptor 1, chemokine XC receptor 1, G protein-coupled receptor 5, GPR5, GPR5chemokine (C motif) XC receptor 1, G-protein coupled receptor 5, Lymphotactin receptor | |
| XCR1 | |
| IgG | |
| Affinity Purified | |
| Detects human XCR1 in direct ELISAs and Western blots. |
| Flow Cytometry 10 uL/10^6 cells | |
| Polyclonal | |
| Goat | |
| P46094 | |
| 2829 | |
| E. coli-derived recombinant human XCR1 Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267 Accession # P46094 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title