missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human XCR1 PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems FAB857P-025
This item is not returnable.
View return policy
Description
XCR1/CCXCR1 Polyclonal specifically detects XCR1/CCXCR1 in Human samples. It is validated for Flow Cytometry.
Specifications
| XCR1/CCXCR1 | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| CCXCR1, CCXCR1XC chemokine receptor 1, chemokine (C motif) receptor 1, chemokine XC receptor 1, G protein-coupled receptor 5, GPR5, GPR5chemokine (C motif) XC receptor 1, G-protein coupled receptor 5, Lymphotactin receptor | |
| Goat | |
| Affinity Purified | |
| Primary | |
| Detects human XCR1 in direct ELISAs and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| R-PE | |
| P46094 | |
| XCR1 | |
| E. coli-derived recombinant human XCR1 Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267 Accession # P46094 | |
| 25 Tests | |
| 2829 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction