missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Testican 1/SPOCK1 Antibody, R&D Systems™

Goat Polyclonal Antibody
£167.00 - £417.00
Specifications
| Antigen | Testican 1/SPOCK1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18726327
![]() |
R&D Systems
AF2327-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18068841
![]() |
R&D Systems
AF2327 |
100 μg |
£417.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Testican 1/SPOCK1 Polyclonal specifically detects Testican 1/SPOCK1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Testican 1/SPOCK1 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| FLJ37170, Protein SPOCK, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1, SPOCK1, SPOCKcwcv and kazal-like domains proteoglycan (testican), TIC1, TICN1 | |
| SPOCK1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q08629 | |
| 6695 | |
| Mouse myeloma cell line NS0-derived recombinant human Testican 1/SPOCK1 Arg22-Trp439 Accession # Q08629 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Testican 1/SPOCK1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) Testican 2 is observed and less than 2% cross-reactivity with rhTestican 3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title