Learn More
Human Testican 1/SPOCK1 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF2327-SP
 View more versions of this product
Product Code. 18726327
  • £167.00 / 25µg
Description
Testican 1/SPOCK1 Polyclonal specifically detects Testican 1/SPOCK1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Testican 1/SPOCK1
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
FLJ37170, Protein SPOCK, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1, SPOCK1, SPOCKcwcv and kazal-like domains proteoglycan (testican), TIC1, TICN1
Goat
Affinity Purified
RUO
6695
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q08629
SPOCK1
Mouse myeloma cell line NS0-derived recombinant human Testican 1/SPOCK1 Arg22-Trp439 Accession # Q08629
25 μg
Primary
Detects human Testican 1/SPOCK1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) Testican 2 is observed and less than 2% cross-reactivity with rhTestican 3 is observed.