missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Testican 1/SPOCK1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2327
This item is not returnable.
View return policy
Description
Testican 1/SPOCK1 Polyclonal specifically detects Testican 1/SPOCK1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Testican 1/SPOCK1 | |
| Polyclonal | |
| Unconjugated | |
| Q08629 | |
| SPOCK1 | |
| Mouse myeloma cell line NS0-derived recombinant human Testican 1/SPOCK1 Arg22-Trp439 Accession # Q08629 | |
| 100 μg | |
| Primary | |
| Detects human Testican 1/SPOCK1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) Testican 2 is observed and less than 2% cross-reactivity with rhTestican 3 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| FLJ37170, Protein SPOCK, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1, SPOCK1, SPOCKcwcv and kazal-like domains proteoglycan (testican), TIC1, TICN1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6695 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction