missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Placental Lactogen/CSH1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£146.00 - £444.00
Specifications
| Antigen | Placental Lactogen/CSH1 |
|---|---|
| Clone | 658234 |
| Dilution | Neutralization 5-50 ng/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18744147
![]() |
R&D Systems
MAB57571-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773057
![]() |
R&D Systems
MAB57571-100 |
100 μg |
£444.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Placental Lactogen/CSH1 Monoclonal antibody specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for NeutralizationSpecifications
| Placental Lactogen/CSH1 | |
| Neutralization 5-50 ng/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH1, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, PL, Placental lactogen, PLchoriomammotropin | |
| Chinese Hamster Ovary cell line CHO-derived human Placental Lactogen/CSH1, Val27-Phe217, Accession # P01243 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 658234 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01243 | |
| 1442 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title