missing translation for 'onlineSavingsMsg'
Learn More

Human Placental Lactogen/CSH1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18744147
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18744147 25 μg 25µg
18773057 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18744147 Supplier R&D Systems Supplier No. MAB57571SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Placental Lactogen/CSH1 Monoclonal antibody specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen Placental Lactogen/CSH1
Applications Neutralization
Classification Monoclonal
Clone 658234
Conjugate Unconjugated
Dilution Neutralization 5-50 ng/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P01243
Gene Alias Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH1, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, PL, Placental lactogen, PLchoriomammotropin
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line CHO-derived human Placental Lactogen/CSH1, Val27-Phe217, Accession # P01243
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1442
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.