missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Placental Lactogen/CSH1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB57571-100
This item is not returnable.
View return policy
Description
Placental Lactogen/CSH1 Monoclonal antibody specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for Neutralization
Specifications
| Placental Lactogen/CSH1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH1, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, PL, Placental lactogen, PLchoriomammotropin | |
| Chinese Hamster Ovary cell line CHO-derived human Placental Lactogen/CSH1, Val27-Phe217, Accession # P01243 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Neutralization | |
| 658234 | |
| Neutralization 5-50 ng/mL | |
| P01243 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1442 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction