missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-MSK1(S376)/MSK2(S360) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£167.00 - £368.00
Specifications
| Antigen | MSK1/MSK2 |
|---|---|
| Clone | 1013F |
| Dilution | Immunocytochemistry 1-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18744309
![]() |
R&D Systems
MAB25181-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18764647
![]() |
R&D Systems
MAB25181 |
100 μg |
£368.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSK1/MSK2 Monoclonal specifically detects MSK1/MSK2 in Human samples. It is validated for Immunocytochemistry.Specifications
| MSK1/MSK2 | |
| Immunocytochemistry 1-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| 1110069D02Rik, 3110005L17Rik, 6330404E13Rik, RPS6KA4, RPS6KA5, RSKB | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1013F | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O75582 | |
| Phosphopeptide containing the human MSK1(S376) site. Accession # O75582 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS | |
| Detects human MSK1 and MSK2 when phosphorylated at S376 and S360, respectively. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title