missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-MSK1(S376)/MSK2(S360) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB25181-SP
This item is not returnable.
View return policy
Description
MSK1/MSK2 Monoclonal specifically detects MSK1/MSK2 in Human samples. It is validated for Immunocytochemistry.
Specifications
| MSK1/MSK2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| 1110069D02Rik, 3110005L17Rik, 6330404E13Rik, RPS6KA4, RPS6KA5, RSKB | |
| Phosphopeptide containing the human MSK1(S376) site. Accession # O75582 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Immunocytochemistry | |
| 1013F | |
| Immunocytochemistry 1-25 ug/mL | |
| O75582 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects human MSK1 and MSK2 when phosphorylated at S376 and S360, respectively. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction