missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-MSK1(S376)/MSK2(S360) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB25181
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
MSK1/MSK2 Monoclonal specifically detects MSK1/MSK2 in Human samples. It is validated for Immunocytochemistry.
Tekniske data
| MSK1/MSK2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| 1110069D02Rik, 3110005L17Rik, 6330404E13Rik, RPS6KA4, RPS6KA5, RSKB | |
| Phosphopeptide containing the human MSK1(S376) site. Accession # O75582 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Immunocytochemistry | |
| 1013F | |
| Immunocytochemistry 1-25 ug/mL | |
| O75582 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects human MSK1 and MSK2 when phosphorylated at S376 and S360, respectively. | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion