missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MIS/AMH Antibody, R&D Systems™

Goat Polyclonal Antibody
£171.00 - £442.00
Specifications
| Antigen | MIS/AMH |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18738767
![]() |
R&D Systems
AF1737-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18030502
|
R&D Systems
AF1737 |
100 μg |
£442.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MIS/AMH Polyclonal specifically detects MIS/AMH in Human samples. It is validated for Western Blot.Specifications
| MIS/AMH | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
| AMH | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q6GTN3 | |
| 268 | |
| E. coli-derived recombinant human MIS/AMH (R&D Systems, Catalog # 1737-MS) Ala453-Arg560 Accession # Q6GTN3 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human MIS/AMH in direct ELISAs and Western blots. In these formats, approximately 40% cross-reactivity with recombinant rat MIS and recombinant mouse MIS is observed. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel