missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MIS/AMH Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1737-SP
This item is not returnable.
View return policy
Description
MIS/AMH Polyclonal specifically detects MIS/AMH in Human samples. It is validated for Western Blot.
Specifications
| MIS/AMH | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 268 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q6GTN3 | |
| AMH | |
| E. coli-derived recombinant human MIS/AMH (R&D Systems, Catalog # 1737-MS) Ala453-Arg560 Accession # Q6GTN3 | |
| 25 μg | |
| Primary | |
| Detects human MIS/AMH in direct ELISAs and Western blots. In these formats, approximately 40% cross-reactivity with recombinant rat MIS and recombinant mouse MIS is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction