missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MIS/AMH Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1737
This item is not returnable.
View return policy
Description
MIS/AMH Polyclonal specifically detects MIS/AMH in Human samples. It is validated for Western Blot.
Specifications
| MIS/AMH | |
| Polyclonal | |
| Unconjugated | |
| Q6GTN3 | |
| AMH | |
| E. coli-derived recombinant human MIS/AMH (R&D Systems, Catalog # 1737-MS) Ala453-Arg560 Accession # Q6GTN3 | |
| 100 μg | |
| Primary | |
| Detects human MIS/AMH in direct ELISAs and Western blots. In these formats, approximately 40% cross-reactivity with recombinant rat MIS and recombinant mouse MIS is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 268 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction