missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kremen-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £345.00
Specifications
| Antigen | Kremen-1 |
|---|---|
| Clone | 301324 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736428
![]() |
R&D Systems
MAB2127-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18162921
|
R&D Systems
MAB2127 |
100 μg |
£345.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kremen-1 Monoclonal specifically detects Kremen-1 in Human samples. It is validated for Western Blot.Specifications
| Kremen-1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_114434 | |
| 83999 | |
| Mouse myeloma cell line NS0-derived recombinant human Kremen-1 Ala20-Gly397 Accession # NP_114434 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Kremen-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Kremen-1 or recombinant human Kremen-2 is observed. |
| 301324 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, FLJ31863, KREMEM1, KREMEN, kremen protein 1, Kremen1, kringle containing transmembrane protein, kringle containing transmembrane protein 1, Kringle domain-containing transmembrane protein 1, kringle-coding gene marking the eye and the nose, Kringle-containing protein marking the eye and the nose, KRM1, KRM1Dickkopf receptor | |
| KREMEN1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title