missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kremen-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2127
This item is not returnable.
View return policy
Description
Kremen-1 Monoclonal specifically detects Kremen-1 in Human samples. It is validated for Western Blot.
Specifications
| Kremen-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_114434 | |
| KREMEN1 | |
| Mouse myeloma cell line NS0-derived recombinant human Kremen-1 Ala20-Gly397 Accession # NP_114434 | |
| 100 μg | |
| Primary | |
| Detects human Kremen-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Kremen-1 or recombinant human Kremen-2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 301324 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, FLJ31863, KREMEM1, KREMEN, kremen protein 1, Kremen1, kringle containing transmembrane protein, kringle containing transmembrane protein 1, Kringle domain-containing transmembrane protein 1, kringle-coding gene marking the eye and the nose, Kringle-containing protein marking the eye and the nose, KRM1, KRM1Dickkopf receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 83999 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction