missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kremen-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2127-SP
This item is not returnable.
View return policy
Description
Kremen-1 Monoclonal specifically detects Kremen-1 in Human samples. It is validated for Western Blot.
Specifications
| Kremen-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_114434 | |
| KREMEN1 | |
| Mouse myeloma cell line NS0-derived recombinant human Kremen-1 Ala20-Gly397 Accession # NP_114434 | |
| 25 μg | |
| Primary | |
| Detects human Kremen-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Kremen-1 or recombinant human Kremen-2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 301324 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, FLJ31863, KREMEM1, KREMEN, kremen protein 1, Kremen1, kringle containing transmembrane protein, kringle containing transmembrane protein 1, Kringle domain-containing transmembrane protein 1, kringle-coding gene marking the eye and the nose, Kringle-containing protein marking the eye and the nose, KRM1, KRM1Dickkopf receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 83999 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido