missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human G6PD Antibody, R&D Systems™

Mouse Monoclonal Antibody
£218.00 - £510.00
Specifications
| Antigen | Glucose 6 Phosphate Dehydrogenase |
|---|---|
| Clone | 1067503 |
| Dilution | Western Blot 1 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 3-25 μg/mL |
| Applications | Western Blot, Simple Western, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18784414
![]() |
R&D Systems
MAB11467-SP |
25 μg |
£218.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18764464
![]() |
R&D Systems
MAB11467-100 |
100 μg |
£510.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, Simple Western, ImmunohistochemistrySpecifications
| Glucose 6 Phosphate Dehydrogenase | |
| Western Blot 1 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| G6PD, G6PD1EC 1.1.1.49, G6PDH, glucose-6-phosphate dehydrogenase | |
| E. coli-derived human G6PD, Ala2-Leu515, Accession # P11413 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 1067503 | |
| Western Blot, Simple Western, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P11413 | |
| 2539 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title