missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human G6PD Antibody, R&D Systems™

Description
Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry
Specifications
Specifications
| Antigen | Glucose 6 Phosphate Dehydrogenase |
| Applications | Western Blot, Simple Western, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 1067503 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 3-25 μg/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | P11413 |
| Gene Alias | G6PD, G6PD1EC 1.1.1.49, G6PDH, glucose-6-phosphate dehydrogenase |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?