missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human G6PD Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB11467-100
This item is not returnable.
View return policy
Description
Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry
Specifications
| Glucose 6 Phosphate Dehydrogenase | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| G6PD, G6PD1EC 1.1.1.49, G6PDH, glucose-6-phosphate dehydrogenase | |
| E. coli-derived human G6PD, Ala2-Leu515, Accession # P11413 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Western Blot, Simple Western, Immunohistochemistry | |
| 1067503 | |
| Western Blot 1 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 3-25 μg/mL | |
| P11413 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2539 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction