missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Epimorphin/Syntaxin 2 Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £345.00
Specifications
| Antigen | Epimorphin/Syntaxin 2 |
|---|---|
| Clone | 338824 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721109
![]() |
R&D Systems
MAB2936-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18102231
|
R&D Systems
MAB2936 |
100 μg |
£345.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Epimorphin/Syntaxin 2 Monoclonal specifically detects Epimorphin/Syntaxin 2 in Human samples. It is validated for Western Blot.Specifications
| Epimorphin/Syntaxin 2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human | |
| P32856 | |
| 2054 | |
| E. coli-derived recombinant human Epimorphin/Syntaxin 2 Met1-Arg188 Accession # P32856 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Epimorphin/Syntaxin 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse Epimorphin is observed. |
| 338824 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EPIMepimorphin, EPM, STX2, STX2AMGC51014, STX2Bsyntaxin-2, STX2CEpimorphin, Syntaxin 2 | |
| STX2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title