missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Epimorphin/Syntaxin 2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2936-SP
This item is not returnable.
View return policy
Description
Epimorphin/Syntaxin 2 Monoclonal specifically detects Epimorphin/Syntaxin 2 in Human samples. It is validated for Western Blot.
Specifications
| Epimorphin/Syntaxin 2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P32856 | |
| STX2 | |
| E. coli-derived recombinant human Epimorphin/Syntaxin 2 Met1-Arg188 Accession # P32856 | |
| 25 μg | |
| Primary | |
| Detects human Epimorphin/Syntaxin 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse Epimorphin is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 338824 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EPIMepimorphin, EPM, STX2, STX2AMGC51014, STX2Bsyntaxin-2, STX2CEpimorphin, Syntaxin 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2054 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction