Learn More
Human Epimorphin/Syntaxin 2 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand:  R&D Systems MAB2936
 View more versions of this product
Product Code. 18102231
  • £345.00 / 100µg
Description
Epimorphin/Syntaxin 2 Monoclonal specifically detects Epimorphin/Syntaxin 2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Epimorphin/Syntaxin 2
Monoclonal
LYOPH
Western Blot 1 ug/mL
P32856
STX2
E. coli-derived recombinant human Epimorphin/Syntaxin 2 Met1-Arg188 Accession # P32856
100 μg
Primary
Detects human Epimorphin/Syntaxin 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse Epimorphin is observed.
Human
Purified
Western Blot
338824
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
EPIMepimorphin, EPM, STX2, STX2AMGC51014, STX2Bsyntaxin-2, STX2CEpimorphin, Syntaxin 2
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
2054
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1