missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chondrolectin Antibody, R&D Systems™

Goat Polyclonal Antibody
£146.00 - £423.00
Specifications
| Antigen | Chondrolectin |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746507
![]() |
R&D Systems
AF2576-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18001412
![]() |
R&D Systems
AF2576 |
100 μg |
£423.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chondrolectin Polyclonal specifically detects Chondrolectin in Human samples. It is validated for Western Blot.Specifications
| Chondrolectin | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| C21orf68, CHODL, chondrolectin, chromosome 21 open reading frame 68, FLJ12627, MT75, PRED12, Transmembrane protein MT75 | |
| CHODL | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9H9P2 | |
| 140578 | |
| Mouse myeloma cell line NS0-derived recombinant human Chondrolectin Arg22-Asn216 Accession # Q9H9P2 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Chondrolectin in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title