missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chondrolectin Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2576-SP
This item is not returnable.
View return policy
Description
Chondrolectin Polyclonal specifically detects Chondrolectin in Human samples. It is validated for Western Blot.
Specifications
| Chondrolectin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C21orf68, CHODL, chondrolectin, chromosome 21 open reading frame 68, FLJ12627, MT75, PRED12, Transmembrane protein MT75 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 140578 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9H9P2 | |
| CHODL | |
| Mouse myeloma cell line NS0-derived recombinant human Chondrolectin Arg22-Asn216 Accession # Q9H9P2 | |
| 25 μg | |
| Primary | |
| Detects human Chondrolectin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction